Learn More
2-Bromo-N,N-diethylethylamine hydrobromide, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 426860250
| Quantity | 25g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C6H15Br2N | |
| MFCD00040376 | |
| 2-diethylamino ethyl bromide hydrobromide, 2-bromo-n,n-diethylethylamine hydrobromide, 2-bromoethyldiethylammonium bromide, 2-bromoethyl diethylamine hydrobromide, 2-bromo-n,n-diethylethanaminium bromide, 2-bromotriethylamine hydrobromide, 2-bromoethyldiethylamine hydrobromide, diethylaminoethyl bromide hydrobromide, 2-bromo-n,n-diethylethanamine hydrobromide, ethanamine, 2-bromo-n,n-diethyl-, hydrobromide | |
| 2-bromo-N,N-diethylethanamine;hydrobromide |
Specifications
| 2-Bromo-N, N-diethylethylamine hydrobromide | |
| 97.5 | |
| Authentic | |
| Glass bottle | |
| (CH3CH2)2N(CH2)2Br·HBr | |
| MFCD00040376 | |
| HLMHCDKXKXBKQK-UHFFFAOYSA-N | |
| 2-bromo-N,N-diethylethanamine;hydrobromide | |
| 70607 | |
| 98% |
| 1069-72-3 | |
| 100.0 | |
| 97.5% min. (Argentometry) | |
| C6H15Br2N | |
| 25g | |
| 2-diethylamino ethyl bromide hydrobromide, 2-bromo-n,n-diethylethylamine hydrobromide, 2-bromoethyldiethylammonium bromide, 2-bromoethyl diethylamine hydrobromide, 2-bromo-n,n-diethylethanaminium bromide, 2-bromotriethylamine hydrobromide, 2-bromoethyldiethylamine hydrobromide, diethylaminoethyl bromide hydrobromide, 2-bromo-n,n-diethylethanamine hydrobromide, ethanamine, 2-bromo-n,n-diethyl-, hydrobromide | |
| [H+].[Br-].CCN(CC)CCBr | |
| 261.00 | |
| 260.99 |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 213-962-3
Promotions