missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-L-phenylalanine, 95%

Catalog No. AAH6314106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6314106 5 g
AAH6314103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6314106 Supplier Thermo Scientific Chemicals Supplier No. H6314106

CAS: 42538-40-9 | C9H10BrNO2 | 244.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42538-40-9
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.09
MDL Number MFCD03092958
InChI Key JFVLNTLXEZDFHW-QMMMGPOBSA-N
Synonym l-2-bromophenylalanine, 2-bromo-l-phenylalanine, s-2-amino-3-2-bromophenyl propanoic acid, 2-bromophenylalanine, 2s-2-amino-3-2-bromophenyl propanoic acid, h-phe 2-br-oh, o-bromo-l-phenylalanine, l-2-br-phe-oh, s-2-bromophenylalanine, l-2-bromophe
PubChem CID 193338
IUPAC Name (2S)-2-amino-3-(2-bromophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=CC=C1Br)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 244.09
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-Bromo-L-phenylalanine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only