missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-9-hexyl-9H-carbazole 98.0+%, TCI America™
$301.98 - $904.06
Chemical Identifiers
| CAS | 864550-95-8 |
|---|---|
| Molecular Formula | C18H20BrN |
| Molecular Weight (g/mol) | 330.269 |
| InChI Key | YGTIVQCXTOGVMV-UHFFFAOYSA-N |
| PubChem CID | 86070322 |
| IUPAC Name | 2-bromo-9-hexylcarbazole |
| SMILES | CCCCCCN1C2=CC=CC=C2C3=C1C=C(C=C3)Br |
Chemical Identifiers
| 864550-95-8 | |
| 330.269 | |
| 86070322 | |
| CCCCCCN1C2=CC=CC=C2C3=C1C=C(C=C3)Br |
| C18H20BrN | |
| YGTIVQCXTOGVMV-UHFFFAOYSA-N | |
| 2-bromo-9-hexylcarbazole |
Specifications
| 864550-95-8 | |
| White-Yellow | |
| 1 g | |
| CCCCCCN1C2=CC=CC=C2C3=C1C=C(C=C3)Br | |
| 330.269 | |
| 330.27 | |
| Crystalline Powder |
| 42°C | |
| C18H20BrN | |
| YGTIVQCXTOGVMV-UHFFFAOYSA-N | |
| 2-bromo-9-hexylcarbazole | |
| 86070322 | |
| ≥98.0% (GC) | |
| 2-Bromo-9-hexyl-9H-carbazole |
Safety and Handling
TSCA : No