missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-9-hexyl-9H-carbazole 98.0+%, TCI America™

Catalog No. B56075G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56075G 5 g
B56071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56075G Supplier TCI America Supplier No. B56075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 864550-95-8
Molecular Formula C18H20BrN
Molecular Weight (g/mol) 330.269
InChI Key YGTIVQCXTOGVMV-UHFFFAOYSA-N
PubChem CID 86070322
IUPAC Name 2-bromo-9-hexylcarbazole
SMILES CCCCCCN1C2=CC=CC=C2C3=C1C=C(C=C3)Br

Specifications

Melting Point 42°C
Color White-Yellow
Quantity 5 g
Formula Weight 330.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-9-hexyl-9H-carbazole