missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-6-(trifluoromethyl)pyridine 98.0+%, TCI America™
$33.56 - $67.11
Chemical Identifiers
| CAS | 189278-27-1 |
|---|---|
| Molecular Formula | C6H3BrF3N |
| Molecular Weight (g/mol) | 226.00 |
| MDL Number | MFCD00153087 |
| InChI Key | DOWNSQADAFSSAR-UHFFFAOYSA-N |
| Synonym | 2-bromo-6-trifluoromethyl pyridine, pyridine, 2-bromo-6-trifluoromethyl, 2-trifluoromethyl-6-bromopyridine, 2-bromo-6-trifluoromethyl-pyridine, 6-bromo-2-trifluoromethyl pyridine, 2-bromo-6-trifluromethylpyridine, 2-bromo-6-trifluormethyl-pyridine, pubchem9307, acmc-209esf, ksc495o4t |
| PubChem CID | 2782902 |
| IUPAC Name | 2-bromo-6-(trifluoromethyl)pyridine |
| SMILES | FC(F)(F)C1=NC(Br)=CC=C1 |
Chemical Identifiers
| 189278-27-1 | |
| 226.00 | |
| DOWNSQADAFSSAR-UHFFFAOYSA-N | |
| 2782902 | |
| FC(F)(F)C1=NC(Br)=CC=C1 |
| C6H3BrF3N | |
| MFCD00153087 | |
| 2-bromo-6-trifluoromethyl pyridine, pyridine, 2-bromo-6-trifluoromethyl, 2-trifluoromethyl-6-bromopyridine, 2-bromo-6-trifluoromethyl-pyridine, 6-bromo-2-trifluoromethyl pyridine, 2-bromo-6-trifluromethylpyridine, 2-bromo-6-trifluormethyl-pyridine, pubchem9307, acmc-209esf, ksc495o4t | |
| 2-bromo-6-(trifluoromethyl)pyridine |
Specifications
| 189278-27-1 | |
| White-Yellow | |
| C6H3BrF3N | |
| 1 g | |
| DOWNSQADAFSSAR-UHFFFAOYSA-N | |
| 2-bromo-6-(trifluoromethyl)pyridine | |
| 2782902 | |
| ≥98.0% (GC) | |
| 2-Bromo-6-(trifluoromethyl)pyridine |
| 50°C | |
| 96°C | |
| MFCD00153087 | |
| 2-bromo-6-trifluoromethyl pyridine, pyridine, 2-bromo-6-trifluoromethyl, 2-trifluoromethyl-6-bromopyridine, 2-bromo-6-trifluoromethyl-pyridine, 6-bromo-2-trifluoromethyl pyridine, 2-bromo-6-trifluromethylpyridine, 2-bromo-6-trifluormethyl-pyridine, pubchem9307, acmc-209esf, ksc495o4t | |
| FC(F)(F)C1=NC(Br)=CC=C1 | |
| 226.00 | |
| 226.00 | |
| Crystalline Powder |
Safety and Handling
TSCA : No