missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-(trifluoromethyl)pyridine 98.0+%, TCI America™

Catalog No. B37591G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37591G 1 g
B37595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37591G Supplier TCI America Supplier No. B37591G
Only null left

Chemical Identifiers

CAS 189278-27-1
Molecular Formula C6H3BrF3N
Molecular Weight (g/mol) 226.00
MDL Number MFCD00153087
InChI Key DOWNSQADAFSSAR-UHFFFAOYSA-N
Synonym 2-bromo-6-trifluoromethyl pyridine, pyridine, 2-bromo-6-trifluoromethyl, 2-trifluoromethyl-6-bromopyridine, 2-bromo-6-trifluoromethyl-pyridine, 6-bromo-2-trifluoromethyl pyridine, 2-bromo-6-trifluromethylpyridine, 2-bromo-6-trifluormethyl-pyridine, pubchem9307, acmc-209esf, ksc495o4t
PubChem CID 2782902
IUPAC Name 2-bromo-6-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=NC(Br)=CC=C1

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 96°C
Quantity 1 g
Formula Weight 226.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-6-(trifluoromethyl)pyridine