missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-(trifluoromethyl)pyridine, 97%

CAS: 189278-27-1 | C6H3BrF3N | 226.00 g/mol

$241.85 - $1032.70

Chemical Identifiers

CAS 189278-27-1
Molecular Formula C6H3BrF3N
Molecular Weight (g/mol) 226.00
MDL Number MFCD00153087
InChI Key DOWNSQADAFSSAR-UHFFFAOYSA-N
Synonym 2-bromo-6-trifluoromethyl pyridine, pyridine, 2-bromo-6-trifluoromethyl, 2-trifluoromethyl-6-bromopyridine, 2-bromo-6-trifluoromethyl-pyridine, 6-bromo-2-trifluoromethyl pyridine, 2-bromo-6-trifluromethylpyridine, 2-bromo-6-trifluormethyl-pyridine, pubchem9307, acmc-209esf, ksc495o4t
PubChem CID 2782902
IUPAC Name 2-bromo-6-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=NC(Br)=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2574903
View Documents
Thermo Scientific Chemicals
H2574903
1 g
Each for $241.85
Only null left
 
AAH2574906
View Documents
Thermo Scientific Chemicals
H2574906
5 g
Each for $1,032.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

189278-27-1
226.00
DOWNSQADAFSSAR-UHFFFAOYSA-N
2782902
FC(F)(F)C1=NC(Br)=CC=C1
C6H3BrF3N
MFCD00153087
2-bromo-6-trifluoromethyl pyridine, pyridine, 2-bromo-6-trifluoromethyl, 2-trifluoromethyl-6-bromopyridine, 2-bromo-6-trifluoromethyl-pyridine, 6-bromo-2-trifluoromethyl pyridine, 2-bromo-6-trifluromethylpyridine, 2-bromo-6-trifluormethyl-pyridine, pubchem9307, acmc-209esf, ksc495o4t
2-bromo-6-(trifluoromethyl)pyridine

Specifications

189278-27-1
78°C to 79°C (14 mmHg)
MFCD00153087
2-bromo-6-trifluoromethyl pyridine, pyridine, 2-bromo-6-trifluoromethyl, 2-trifluoromethyl-6-bromopyridine, 2-bromo-6-trifluoromethyl-pyridine, 6-bromo-2-trifluoromethyl pyridine, 2-bromo-6-trifluromethylpyridine, 2-bromo-6-trifluormethyl-pyridine, pubchem9307, acmc-209esf, ksc495o4t
FC(F)(F)C1=NC(Br)=CC=C1
226.00
226
2-Bromo-6-(trifluoromethyl)pyridine
48°C to 51°C
C6H3BrF3N
1 g
DOWNSQADAFSSAR-UHFFFAOYSA-N
2-bromo-6-(trifluoromethyl)pyridine
2782902
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only