missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-pyridinemethanol, 96%

Catalog No. p-7037400
Change view
Click to view available options
Quantity:
1 g
250 mg
5 g
Catalog No. Quantity
AAH56123MD 250 mg
AAH5612303 1 g
AAH5612306 5 g
3 options

Catalog No. AAH56123MD

Supplier: Thermo Scientific Chemicals H56123MD

Only null left

CAS: 33674-96-3 | C6H6BrNO | 188.02 g/mol

It is an active pharmaceutical ingredient and an pharmaceutical adjuvant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical ingredient and an pharmaceutical adjuvant.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents and bases. Store at room temperature.

Chemical Identifiers

CAS 33674-96-3
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
MDL Number MFCD04039884
InChI Key XDDGKNRSCDEWBR-UHFFFAOYSA-N
Synonym 6-bromopyridin-2-yl methanol, 2-bromo-6-pyridinemethanol, 2-bromo-6-hydroxymethyl pyridine, 6-bromo-2-hydroxymethyl pyridine, 2-pyridinemethanol, 6-bromo, 6-bromopyridine-2-methanol, 6-bromo-pyridine-2-yl methanol, 6-bromo-2-pyridinemethanol, 6-bromo-pyridin-2-yl-methanol, 6-bromo-pyridin-2-yl methanol
PubChem CID 3517811
IUPAC Name (6-bromopyridin-2-yl)methanol
SMILES OCC1=CC=CC(Br)=N1

Specifications

Melting Point 34°C to 39°C
Boiling Point 246°C
Flash Point >110°C (230°F)
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 188.02
Percent Purity 96%
Chemical Name or Material 2-Bromo-6-pyridinemethanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only