missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-piperidinopyridine, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC38110EA

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC38110EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

24255-97-8
241.132
YHQZMCNKHWPFBS-UHFFFAOYSA-N
7164597
C1CCN(CC1)C2=NC(=CC=C2)Br
C10H13BrN2
MFCD07772823
2-bromo-6-piperidin-1-yl pyridine, 2-bromo-6-piperidinopyridine, acmc-1cmoy, 6-bromo-2-piperidylpyridine, 2-bromo-6-1-piperidinyl pyridine, 2-bromo-6-piperadinopyridine, 2-bromanyl-6-piperidin-1-yl-pyridine, pyridine,2-bromo-6-1-piperidinyl, 6'-bromo-3,4,5,6-tetrahydro-2h-1,2' bipyridinyl
2-bromo-6-piperidin-1-ylpyridine

Specifications

2-Bromo-6-piperidinopyridine
96.5
97%
C10H13BrN2
MFCD07772823
YHQZMCNKHWPFBS-UHFFFAOYSA-N
2-bromo-6-piperidin-1-ylpyridine
7164597
97%
24255-97-8
100.0
Amber Glass Bottle
10g
2-bromo-6-piperidin-1-yl pyridine, 2-bromo-6-piperidinopyridine, acmc-1cmoy, 6-bromo-2-piperidylpyridine, 2-bromo-6-1-piperidinyl pyridine, 2-bromo-6-piperadinopyridine, 2-bromanyl-6-piperidin-1-yl-pyridine, pyridine,2-bromo-6-1-piperidinyl, 6'-bromo-3,4,5,6-tetrahydro-2h-1,2' bipyridinyl
C1CCN(CC1)C2=NC(=CC=C2)Br
241.132
241.13
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents