missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-phenylpyridine 98.0+%, TCI America™

$88.56 - $263.75

Chemical Identifiers

CAS 39774-26-0
Molecular Formula C11H8BrN
Molecular Weight (g/mol) 234.096
MDL Number MFCD00234691
InChI Key XIYPPJVLAAXYAB-UHFFFAOYSA-N
PubChem CID 568679
IUPAC Name 2-bromo-6-phenylpyridine
SMILES C1=CC=C(C=C1)C2=NC(=CC=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B5150200MG
View Documents
TCI America
B5150200MG
200 mg
Each for $88.56
Only null left
 
B51501G
View Documents
TCI America
B51501G
1 g
Each for $263.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

39774-26-0
234.096
XIYPPJVLAAXYAB-UHFFFAOYSA-N
2-bromo-6-phenylpyridine
C11H8BrN
MFCD00234691
568679
C1=CC=C(C=C1)C2=NC(=CC=C2)Br

Specifications

39774-26-0
Yellow
MFCD00234691
XIYPPJVLAAXYAB-UHFFFAOYSA-N
2-bromo-6-phenylpyridine
568679
≥98.0% (GC)
2-Bromo-6-phenylpyridine
52°C
C11H8BrN
200 mg
C1=CC=C(C=C1)C2=NC(=CC=C2)Br
234.096
234.10
Crystalline Lumps
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents