missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-6-methylpyridine 98.0+%, TCI America™
Supplier: TCI America B211425G
Specifications
| 2-Bromo-6-methylpyridine | |
| Yellow | |
| C6H6BrN | |
| 25 g | |
| SOHDPICLICFSOP-UHFFFAOYSA-N | |
| 2-bromo-6-methylpyridine | |
| 79205 | |
| ≥98.0% (GC,T) |
| 5315-25-3 | |
| 102°C | |
| MFCD00040743 | |
| 2-bromo-6-picoline, 6-bromo-2-picoline, 6-bromo-2-methylpyridine, 2-bromo-6-methyl-pyridine, 2-bromo-6-methyl pyridine, 2-brom-6-methylpyridin, pyridine, 2-bromo-6-methyl, 2-bromo-6-methy pyridine, 6-bromopicoline, 6-methyl-2-bromopyridine | |
| CC1=CC=CC(Br)=N1 | |
| 172.03 | |
| 172.03 | |
| Liquid |
Chemical Identifiers
| 5315-25-3 | |
| 172.03 | |
| SOHDPICLICFSOP-UHFFFAOYSA-N | |
| 79205 | |
| CC1=CC=CC(Br)=N1 |
| C6H6BrN | |
| MFCD00040743 | |
| 2-bromo-6-picoline, 6-bromo-2-picoline, 6-bromo-2-methylpyridine, 2-bromo-6-methyl-pyridine, 2-bromo-6-methyl pyridine, 2-brom-6-methylpyridin, pyridine, 2-bromo-6-methyl, 2-bromo-6-methy pyridine, 6-bromopicoline, 6-methyl-2-bromopyridine | |
| 2-bromo-6-methylpyridine |
Safety and Handling
TSCA : No