missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-6-methoxypyridine 97.0+%, TCI America™
Supplier: TCI America B326025G
Specifications
| 2-Bromo-6-methoxypyridine | |
| Yellow | |
| C6H6BrNO | |
| 25 g | |
| KMODISUYWZPVGV-UHFFFAOYSA-N | |
| 2-bromo-6-methoxypyridine | |
| 256810 | |
| ≥97.0% (GC) |
| 40473-07-2 | |
| 91°C | |
| MFCD00088345 | |
| 2-bromo-6-methoxy-pyridine, pyridine, 2-bromo-6-methoxy, 6-bromo-2-methoxypyridine, 2-methoxy-6-bromo pyridine, buttpark 82\06-06, 2-brom-6-methoxypyridin, zlchem 781, pubchem9206, acmc-209jec, 2-bromo-6-methoxy-pyridin | |
| COC1=CC=CC(Br)=N1 | |
| 188.02 | |
| 188.02 | |
| Liquid |
Chemical Identifiers
| 40473-07-2 | |
| 188.02 | |
| KMODISUYWZPVGV-UHFFFAOYSA-N | |
| 256810 | |
| COC1=CC=CC(Br)=N1 |
| C6H6BrNO | |
| MFCD00088345 | |
| 2-bromo-6-methoxy-pyridine, pyridine, 2-bromo-6-methoxy, 6-bromo-2-methoxypyridine, 2-methoxy-6-bromo pyridine, buttpark 82\06-06, 2-brom-6-methoxypyridin, zlchem 781, pubchem9206, acmc-209jec, 2-bromo-6-methoxy-pyridin | |
| 2-bromo-6-methoxypyridine |
Safety and Handling
TSCA : No