missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-methoxypyridine, 97%

Catalog No. AAH3059306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3059306 5 g
AAH3059314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3059306 Supplier Thermo Scientific Chemicals Supplier No. H3059306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 40473-07-2 | C6H6BrNO | 188.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40473-07-2
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
MDL Number MFCD00088345
InChI Key KMODISUYWZPVGV-UHFFFAOYSA-N
Synonym 2-bromo-6-methoxy-pyridine, pyridine, 2-bromo-6-methoxy, 6-bromo-2-methoxypyridine, 2-methoxy-6-bromo pyridine, buttpark 82\06-06, 2-brom-6-methoxypyridin, zlchem 781, pubchem9206, acmc-209jec, 2-bromo-6-methoxy-pyridin
PubChem CID 256810
IUPAC Name 2-bromo-6-methoxypyridine
SMILES COC1=CC=CC(Br)=N1

Specifications

Density 1.53
Boiling Point 206°C
Flash Point 104°C (219°F)
Refractive Index 1.56
Quantity 5 g
Formula Weight 188.02
Percent Purity 97%
Chemical Name or Material 2-Bromo-6-methoxypyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only