missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-methoxynaphthalene 98.0+%, TCI America™

Catalog No. p-7119111
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B154725G 25 g
B1547250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B154725G Supplier TCI America Supplier No. B154725G

Chemical Identifiers

CAS 5111-65-9
Molecular Formula C11H9BrO
Molecular Weight (g/mol) 237.096
MDL Number MFCD00004062
InChI Key AYFJBMBVXWNYLT-UHFFFAOYSA-N
Synonym 6-methoxy-2-bromonaphthalene, naphthalene, 2-bromo-6-methoxy, 6-bromo-2-methoxynaphthalene, 2-bromo-6-methoxy-naphthalene, 6-brom-2-naphthyl-methyl ether, 2-bromo-6-methoxynapthalene, unii-s19t080mrg, naproxen ep impurity n, bromo 2--6-methoxy naphthalene, 2-methoxy-6-bromonaphthalene
PubChem CID 78786
IUPAC Name 2-bromo-6-methoxynaphthalene
SMILES COC1=CC2=C(C=C1)C=C(C=C2)Br

Specifications

Melting Point 108°C
Color White-Yellow
Boiling Point 164°C
Quantity 25 g
Formula Weight 237.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-6-methoxynaphthalene