missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-methoxynaphthalene, 98%

Catalog No. AAA1945009
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1945009 10 g
AAA1945018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1945009 Supplier Thermo Scientific Chemicals Supplier No. A1945009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5111-65-9 | C11H9BrO | 237.096 g/mol

2-Bromo-6-methoxynaphthalene is used in the synthesis of nabumetone [4-(6-methoxy-2-naphthalenyl)-2-butanone] by Heck reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-6-methoxynaphthalene is used in the synthesis of nabumetone [4-(6-methoxy-2-naphthalenyl)-2-butanone] by Heck reaction.

Solubility
Soluble in DMSO. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 5111-65-9
Molecular Formula C11H9BrO
Molecular Weight (g/mol) 237.096
MDL Number MFCD00004062
InChI Key AYFJBMBVXWNYLT-UHFFFAOYSA-N
Synonym 6-methoxy-2-bromonaphthalene, naphthalene, 2-bromo-6-methoxy, 6-bromo-2-methoxynaphthalene, 2-bromo-6-methoxy-naphthalene, 6-brom-2-naphthyl-methyl ether, 2-bromo-6-methoxynapthalene, unii-s19t080mrg, naproxen ep impurity n, bromo 2--6-methoxy naphthalene, 2-methoxy-6-bromonaphthalene
PubChem CID 78786
IUPAC Name 2-bromo-6-methoxynaphthalene
SMILES COC1=CC2=C(C=C1)C=C(C=C2)Br

Specifications

Melting Point 105°C to 110°C
Boiling Point 160°C to 164°C (3 mmHg)
Quantity 10 g
Beilstein 2043874
Solubility Information Soluble in DMSO. Insoluble in water.
Formula Weight 237.1
Percent Purity 98%
Chemical Name or Material 2-Bromo-6-methoxynaphthalene
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 225-837-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only