missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-fluoropyridine 98.0+%, TCI America™

Catalog No. B36671G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36671G 1 g
B36675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36671G Supplier TCI America Supplier No. B36671G

Chemical Identifiers

CAS 144100-07-2
Molecular Formula C5H3BrFN
Molecular Weight (g/mol) 175.988
MDL Number MFCD03095258
InChI Key ZIDIKYIZXMYHAW-UHFFFAOYSA-N
Synonym 6-bromo-2-fluoropyridine, 2-fluoro-6-bromopyridine, 2-bromo-6-fluoro-pyridine, pyridine, 2-bromo-6-fluoro, abbypharma ap-17-5098, acmc-1cab6, 2-bromo-6-fluoro pyridine, ksc485m5f, platinum 1+ , 2-bromo-5-fluoropyridine-n 9-hydroxy-6-methylpyrido 2,3-a pyrrolo 3,4-c carbazole-5,7 1h,6h-dionato-n1,n12 6-methyl-3-pyridinol-n1-, sp-4-2-, hexafluorophosphate 1-
PubChem CID 639438
IUPAC Name 2-bromo-6-fluoropyridine
SMILES C1=CC(=NC(=C1)Br)F

Specifications

Melting Point 32°C
Color White-Yellow
Boiling Point 164°C
Quantity 1 g
Formula Weight 175.99
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromo-6-fluoropyridine