missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-fluoropyridine, 97%

Catalog No. AAH3092403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3092403 1 g
AAH3092406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3092403 Supplier Thermo Scientific Chemicals Supplier No. H3092403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 144100-07-2 | C5H3BrFN | 175.988 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 144100-07-2
Molecular Formula C5H3BrFN
Molecular Weight (g/mol) 175.988
MDL Number MFCD03095258
InChI Key ZIDIKYIZXMYHAW-UHFFFAOYSA-N
Synonym 6-bromo-2-fluoropyridine, 2-fluoro-6-bromopyridine, 2-bromo-6-fluoro-pyridine, pyridine, 2-bromo-6-fluoro, abbypharma ap-17-5098, acmc-1cab6, 2-bromo-6-fluoro pyridine, ksc485m5f, platinum 1+ , 2-bromo-5-fluoropyridine-n 9-hydroxy-6-methylpyrido 2,3-a pyrrolo 3,4-c carbazole-5,7 1h,6h-dionato-n1,n12 6-methyl-3-pyridinol-n1-, sp-4-2-, hexafluorophosphate 1-
PubChem CID 639438
IUPAC Name 2-bromo-6-fluoropyridine
SMILES C1=CC(=NC(=C1)Br)F

Specifications

Melting Point 30°C to 34°C
Quantity 1 g
UN Number UN2811
Formula Weight 175.99
Percent Purity 97%
Chemical Name or Material 2-Bromo-6-fluoropyridine
Hazard Category H301+H311-H315-H319-H332-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only