missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-fluoronaphthalene 97.0+%, TCI America™

Catalog No. B52291G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B52291G Supplier TCI America Supplier No. B52291G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 324-41-4
Molecular Formula C10H6BrF
Molecular Weight (g/mol) 225.06
MDL Number MFCD09763683
InChI Key CAJAZWLCCNCVEY-UHFFFAOYSA-N
PubChem CID 345397
IUPAC Name 2-bromo-6-fluoronaphthalene
SMILES FC1=CC=C2C=C(Br)C=CC2=C1

Specifications

Melting Point 66°C
Boiling Point 140°C
Quantity 1 g
Formula Weight 225.06
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-6-fluoronaphthalene