missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-chlorotoluene 98.0+%, TCI America™

Catalog No. p-7119437
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20325G 5 g
B203225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20325G Supplier TCI America Supplier No. B20325G

Chemical Identifiers

CAS 62356-27-8
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.48
MDL Number MFCD00061121
InChI Key DMARBQGIQKLIPM-UHFFFAOYSA-N
Synonym 2-bromo-6-chlorotoluene, benzene, 1-bromo-3-chloro-2-methyl, pubchem3601, 2-chloro-6-bromotoluene, 6-bromo-2-chlorotoluene, acmc-209n2m, ksc357m2t, attercop-chm at132933, #, 1-bromanyl-3-chloranyl-2-methyl-benzene
PubChem CID 123552
IUPAC Name 1-bromo-3-chloro-2-methylbenzene
SMILES CC1=C(Cl)C=CC=C1Br

Specifications

Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 120°C
Formula Weight 205.48
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Bromo-6-chlorotoluene