missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-(4-methyl-1-piperazinyl)benzonitrile, 95%, Thermo Scientific Chemicals

Catalog No. AAH61858MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61858MD 250 mg
AAH6185803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH61858MD Supplier Thermo Scientific Chemicals Supplier No. H61858MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1260763-01-6 | Molecular Formula: C12H14BrN3 | Molecular Weight: 280.169 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1260763-01-6
Molecular Formula C12H14BrN3
Molecular Weight (g/mol) 280.169
InChI Key XDFWUVBVNHLPIB-UHFFFAOYSA-N
Synonym 2-bromo-6-4-methylpiperazin-1-yl benzonitrile
PubChem CID 75360874
IUPAC Name 2-bromo-6-(4-methylpiperazin-1-yl)benzonitrile
SMILES CN1CCN(CC1)C2=C(C(=CC=C2)Br)C#N

Specifications

Quantity 250 mg
UN Number UN3439
Formula Weight 280.17
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Bromo-6-(4-methyl-1-piperazinyl)benzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only