missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™

Catalog No. p-7123937
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B50771G 1 g
B50775G 5 g
2 options

Catalog No. B50771G

Supplier: TCI America B50771G

Only null left

Chemical Identifiers

CAS 651358-83-7
Molecular Formula C11H15BBrNO2
Molecular Weight (g/mol) 283.96
MDL Number MFCD06796177
InChI Key OAVRLCKBKDMGGW-UHFFFAOYSA-N
Synonym 6-bromopyridine-2-boronic acid pinacol ester, 2-bromo-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-bromo-6-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-bromopyridine-6-boronic acid pinacol ester, 2-6-bromo-2-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, pyridine, 2-bromo-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-bromo-2-pyridylboronic acid pinacol ester, 6-bromo-2-pyridineboronic acid pinacol ester, 6-bromopyridin-2-ylboronic acid pinacol ester, 6-bromopyridin-2-yl boronic acid pinacol ester
PubChem CID 12177444
IUPAC Name 2-bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=NC(=CC=C2)Br

Specifications

Melting Point 132°C
Color White
Quantity 1 g
Formula Weight 283.96
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine