missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 132°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 283.96 |
| Percent Purity | ≥98.0% (GC,T) |
| Grade | Laboratory |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
Safety and Handling
| TSCA | No |