missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-(trimethylsilyl)thiophene 98.0+%, TCI America™

Catalog No. B48551G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B48551G Supplier TCI America Supplier No. B48551G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 18246-28-1
Color Yellow
Boiling Point 70°C
Molecular Formula C7H11BrSSi
Quantity 1 g
Synonym (5-Bromo-2-thienyl)trimethylsilane
InChI Key IXLYRDOGOXBECE-UHFFFAOYSA-N
SMILES C[Si](C)(C)C1=CC=C(S1)Br
IUPAC Name (5-bromothiophen-2-yl)-trimethylsilane
Molecular Weight (g/mol) 235.214
PubChem CID 13201448
Formula Weight 235.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-(trimethylsilyl)thiophene
Show More Show Less