missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitrothiazole 97.0+%, TCI America™

Catalog No. B44131G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44131G 1 g
B44135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44131G Supplier TCI America Supplier No. B44131G

Chemical Identifiers

CAS 3034-48-8
Molecular Formula C3HBrN2O2S
Molecular Weight (g/mol) 209.02
MDL Number MFCD00005317
InChI Key ANIJFZVZXZQFDH-UHFFFAOYSA-N
Synonym 2-bromo-5-nitrothiazole, thiazole, 2-bromo-5-nitro, 5-nitro-2-bromothiazole, pubchem11070, acmc-1ad6f, 2-bromo-5-nitro-thiazole, 4-27-00-00963 beilstein handbook reference, 2-bromo-5-nitrothiozole, #
PubChem CID 18211
IUPAC Name 2-bromo-5-nitro-1,3-thiazole
SMILES [O-][N+](=O)C1=CN=C(Br)S1

Specifications

Melting Point 85°C
Quantity 1 g
Formula Weight 209.02
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-nitrothiazole
RTECSNumber XJ2986000
TSCA Yes