missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitrothiazole, 98%

Catalog No. p-7022092
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1470106 5 g
AAA1470114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1470106 Supplier Thermo Scientific Chemicals Supplier No. A1470106

CAS: 3034-48-8 | C3HBrN2O2S | 209.02 g/mol

2-Bromo-5-nitrothiazole is used as a building block. It is used as an intermediate to produce other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-5-nitrothiazole is used as a building block. It is used as an intermediate to produce other chemicals.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3034-48-8
Molecular Formula C3HBrN2O2S
Molecular Weight (g/mol) 209.02
MDL Number MFCD00005317
InChI Key ANIJFZVZXZQFDH-UHFFFAOYSA-N
Synonym 2-bromo-5-nitrothiazole, thiazole, 2-bromo-5-nitro, 5-nitro-2-bromothiazole, pubchem11070, acmc-1ad6f, 2-bromo-5-nitro-thiazole, 4-27-00-00963 beilstein handbook reference, 2-bromo-5-nitrothiozole, #
PubChem CID 18211
IUPAC Name 2-bromo-5-nitro-1,3-thiazole
SMILES [O-][N+](=O)C1=CN=C(Br)S1

Specifications

Melting Point 85°C to 90°C
Quantity 5 g
Beilstein 4856
Solubility Information Slightly soluble in water.
Formula Weight 209.03
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-nitrothiazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-226-8
RTECSNumber XJ2986000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only