missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-nitropyridine 98.0+%, TCI America™
$44.73 - $123.03
Chemical Identifiers
| CAS | 4487-59-6 |
|---|---|
| Molecular Formula | C5H3BrN2O2 |
| Molecular Weight (g/mol) | 203.00 |
| MDL Number | MFCD00006222 |
| InChI Key | HUUFTVUBFFESEN-UHFFFAOYSA-N |
| Synonym | 2-bromo-5-nitro pyridine, 2-bromo-5-nitro-pyridine, 5-nitro-2-bromopyridine, 2-brom-5-nitropyridin, pubchem1183, acmc-1agnm, 2-bromo-5-nitro-pyridin, ksc237s9f, tpc-py095 |
| PubChem CID | 78240 |
| IUPAC Name | 2-bromo-5-nitropyridine |
| SMILES | [O-][N+](=O)C1=CC=C(Br)N=C1 |
Chemical Identifiers
| 4487-59-6 | |
| 203.00 | |
| HUUFTVUBFFESEN-UHFFFAOYSA-N | |
| 78240 | |
| [O-][N+](=O)C1=CC=C(Br)N=C1 |
| C5H3BrN2O2 | |
| MFCD00006222 | |
| 2-bromo-5-nitro pyridine, 2-bromo-5-nitro-pyridine, 5-nitro-2-bromopyridine, 2-brom-5-nitropyridin, pubchem1183, acmc-1agnm, 2-bromo-5-nitro-pyridin, ksc237s9f, tpc-py095 | |
| 2-bromo-5-nitropyridine |
Specifications
| 4487-59-6 | |
| White-Yellow | |
| C5H3BrN2O2 | |
| 1 g | |
| HUUFTVUBFFESEN-UHFFFAOYSA-N | |
| 2-bromo-5-nitropyridine | |
| 78240 | |
| ≥98.0% (GC) | |
| 2-Bromo-5-nitropyridine |
| 139°C | |
| 147°C | |
| MFCD00006222 | |
| 2-bromo-5-nitro pyridine, 2-bromo-5-nitro-pyridine, 5-nitro-2-bromopyridine, 2-brom-5-nitropyridin, pubchem1183, acmc-1agnm, 2-bromo-5-nitro-pyridin, ksc237s9f, tpc-py095 | |
| [O-][N+](=O)C1=CC=C(Br)N=C1 | |
| 203.00 | |
| 203.00 | |
| Crystalline Powder |
Safety and Handling
TSCA : No