Learn More
2-Bromo-5-methylthiophene, stabilised with Copper (0.1%) and Sodium bicarbonate (0.4, Thermo Scientific™
Chemical Identifiers
| CAS | 765-58-2 |
|---|---|
| Molecular Formula | C5H5BrS |
| Molecular Weight (g/mol) | 177.06 |
| MDL Number | MFCD00130103 |
| InChI Key | ACDLOOGOFKSUPO-UHFFFAOYSA-N |
| Synonym | 2-bromo-5-methyl-thiophene, 2-bromo-5-methyl thiophene, thiophene, 2-bromo-5-methyl, 2-methyl-5-bromothiophene, 2-brom-5-methylthiophen, zlchem 358, pubchem5193, acmc-1beki, 2-methyl-5-bromothiphene, 5-bromo-2-methylthiophene |
| PubChem CID | 69831 |
| IUPAC Name | 2-bromo-5-methylthiophene |
| SMILES | CC1=CC=C(S1)Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC434200010
|
Thermo Scientific Chemicals
434200010 |
1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC434200050
|
Thermo Scientific Chemicals
434200050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 765-58-2 | |
| 177.06 | |
| ACDLOOGOFKSUPO-UHFFFAOYSA-N | |
| 69831 | |
| CC1=CC=C(S1)Br |
| C5H5BrS | |
| MFCD00130103 | |
| 2-bromo-5-methyl-thiophene, 2-bromo-5-methyl thiophene, thiophene, 2-bromo-5-methyl, 2-methyl-5-bromothiophene, 2-brom-5-methylthiophen, zlchem 358, pubchem5193, acmc-1beki, 2-methyl-5-bromothiphene, 5-bromo-2-methylthiophene | |
| 2-bromo-5-methylthiophene |
Specifications
| 765-58-2 | |
| 100.0 | |
| 78°C | |
| 94% min. (GC) | |
| C5H5BrS | |
| 1g | |
| ACDLOOGOFKSUPO-UHFFFAOYSA-N | |
| 2-bromo-5-methylthiophene | |
| stabilizer - Cu (0.1%), NaHCO3 (0.4%) | |
| 177.06 | |
| 2-Bromo-5-methylthiophene, stabilised with Copper (0.1%) and Sodium bicarbonate (0.4%) |
| 94.0 | |
| 65°C (15.0mmHg) | |
| Authentic | |
| Glass bottle | |
| MFCD00130103 | |
| 2-bromo-5-methyl-thiophene, 2-bromo-5-methyl thiophene, thiophene, 2-bromo-5-methyl, 2-methyl-5-bromothiophene, 2-brom-5-methylthiophen, zlchem 358, pubchem5193, acmc-1beki, 2-methyl-5-bromothiphene, 5-bromo-2-methylthiophene | |
| CC1=CC=C(S1)Br | |
| 177.06 | |
| 69831 | |
| ≥94.0% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Warning
EINECSNumber : 212-151-1
RUO â Research Use Only