missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methylthiophene, stabilised with Copper (0.1%) and Sodium bicarbonate (0.4%), 95%

Catalog No. AC434200010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434200010 1 g Glass bottle
AC434200050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434200010 Supplier Thermo Scientific Chemicals Supplier No. 434200010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 765-58-2 | C5H5BrS | 177.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 765-58-2
Molecular Formula C5H5BrS
Molecular Weight (g/mol) 177.06
MDL Number MFCD00130103
InChI Key ACDLOOGOFKSUPO-UHFFFAOYSA-N
Synonym 2-bromo-5-methyl-thiophene, 2-bromo-5-methyl thiophene, thiophene, 2-bromo-5-methyl, 2-methyl-5-bromothiophene, 2-brom-5-methylthiophen, zlchem 358, pubchem5193, acmc-1beki, 2-methyl-5-bromothiphene, 5-bromo-2-methylthiophene
PubChem CID 69831
IUPAC Name 2-bromo-5-methylthiophene
SMILES CC1=CC=C(S1)Br

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Boiling Point 65°C (15.0 mmHg)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Stabilizer stabilizer - Cu (0.1%), NaHCO3 (0.4%)
Formula Weight 177.06
Percent Purity ≥94.0%
Physical Form Liquid, May Include Precipitate
Chemical Name or Material 2-Bromo-5-methylthiophene, stabilised with Copper (0.1%) and Sodium bicarbonate (0.4%)
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
EINECSNumber 212-151-1

RUO – Research Use Only