missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methylthiazole 98.0+%, TCI America™

Catalog No. B46045G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46045G 5 g
B46041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46045G Supplier TCI America Supplier No. B46045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41731-23-1
Molecular Formula C4H4BrNS
Molecular Weight (g/mol) 178.047
MDL Number MFCD08460610
InChI Key FJPZHYAYNAUKKA-UHFFFAOYSA-N
Synonym 2-bromo-5-methylthiazole, 2-bromo-5-methyl-thiazole, thiazole, 2-bromo-5-methyl, pubchem20436, acmc-209jlg, 2-bromo-5-methyl thiazole, ksc493s9j
PubChem CID 21906106
IUPAC Name 2-bromo-5-methyl-1,3-thiazole
SMILES CC1=CN=C(S1)Br

Specifications

Color Yellow
Boiling Point 84°C
Quantity 5 g
Formula Weight 178.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-methylthiazole