missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methylpyrimidine, 95%

CAS: 150010-20-1 | C5H5BrN2 | 173.013 g/mol

$201.25 - $654.93

Chemical Identifiers

CAS 150010-20-1
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.013
MDL Number MFCD09753737
InChI Key KYCGEJNZMHUBMX-UHFFFAOYSA-N
Synonym pyrimidine, 2-bromo-5-methyl, acmc-20a0od, 2-bromo-5-methyl-pyrimidine, 2-?bromo-?5-?methylpyrimidine
PubChem CID 45789625
IUPAC Name 2-bromo-5-methylpyrimidine
SMILES CC1=CN=C(N=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66877MD
View Documents
Thermo Scientific Chemicals
H66877MD
250 mg
Each for $201.25
Only null left
 
AAH6687703
View Documents
Thermo Scientific Chemicals
H6687703
1 g
Each for $654.93
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

150010-20-1
173.013
KYCGEJNZMHUBMX-UHFFFAOYSA-N
45789625
CC1=CN=C(N=C1)Br
C5H5BrN2
MFCD09753737
pyrimidine, 2-bromo-5-methyl, acmc-20a0od, 2-bromo-5-methyl-pyrimidine, 2-?bromo-?5-?methylpyrimidine
2-bromo-5-methylpyrimidine

Specifications

150010-20-1
MFCD09753737
21248671
KYCGEJNZMHUBMX-UHFFFAOYSA-N
2-bromo-5-methylpyrimidine
45789625
95%
2-Bromo-5-methylpyrimidine
C5H5BrN2
250 mg
pyrimidine, 2-bromo-5-methyl, acmc-20a0od, 2-bromo-5-methyl-pyrimidine, 2-?bromo-?5-?methylpyrimidine
CC1=CN=C(N=C1)Br
173.013
173.01
Solid
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only