missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methylpyridine, 98+%

Catalog No. AAB2508306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2508306 5 g
AAB2508314 25 g
AAB2508322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2508306 Supplier Thermo Scientific Chemicals Supplier No. B2508306

CAS: 3510-66-5 | C6H6BrN | 172.025 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3510-66-5
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.025
MDL Number MFCD00209553
InChI Key YWNJQQNBJQUKME-UHFFFAOYSA-N
Synonym 2-bromo-5-picoline, 6-bromo-3-picoline, 2-bromo-5-methyl-pyridine, pyridine, 2-bromo-5-methyl, 2-bromo-5-methyl pyridine, 5-methyl-2-bromopyridine, 2-brom-5-methylpyridin, pubchem1185
PubChem CID 564216
IUPAC Name 2-bromo-5-methylpyridine
SMILES CC1=CN=C(C=C1)Br

Specifications

Melting Point 40°C to 45°C
Boiling Point 95°C to 96°C (12 mmHg)
Flash Point 103°C (217°F)
Quantity 5 g
Beilstein 107323
Formula Weight 172.03
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-5-methylpyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only