missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-iodopyridine 97.0+%, TCI America™

Catalog No. B37655G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37655G 5 g
B376525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37655G Supplier TCI America Supplier No. B37655G

Chemical Identifiers

CAS 73290-22-9
Molecular Formula C5H3BrIN
Molecular Weight (g/mol) 283.89
MDL Number MFCD03095201
InChI Key LLKRSJVPTKFSLS-UHFFFAOYSA-N
Synonym 2-bromo-5-iodo-pyridine, pyridine, 2-bromo-5-iodo, 5-iodo-2-bromopyridine, pubchem5969, 5-iodo-2-bromo-pyridine, acmc-1bb8d, ksc493q0p, 2-bromo-5-iodopyridine
PubChem CID 4738271
IUPAC Name 2-bromo-5-iodopyridine
SMILES BrC1=CC=C(I)C=N1

Specifications

Melting Point 123°C
Color White-Yellow
Quantity 5 g
Formula Weight 283.89
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-iodopyridine