missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-iodo-3-methylpyridine 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | 46°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Synonym | 2-bromo-5-iodo-3-picoline, pyridine, 2-bromo-5-iodo-3-methyl, 2-bromo-5-iodo-3-methyl-pyridine, pubchem5968, acmc-1b7x6, pyridine,2-bromo-5-iodo-3-methyl, 2-bromo-5-iodo-3-picoline; 2-bromo-3-methyl-5-iodopyridine |
| Formula Weight | 297.92 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Bromo-5-iodo-3-methylpyridine |
Safety and Handling
| TSCA | No |