missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-iodo-3-methylpyridine 96.0+%, TCI America™

Catalog No. B43121G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43121G 1 g
B43125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43121G Supplier TCI America Supplier No. B43121G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 65550-78-9 | C6H5BrIN | 297.921 g/mol

Chemical Identifiers

CAS 65550-78-9
Molecular Formula C6H5BrIN
Molecular Weight (g/mol) 297.921
MDL Number MFCD09037466
InChI Key WJPSANINUBHCFE-UHFFFAOYSA-N
PubChem CID 10108680
IUPAC Name 2-bromo-5-iodo-3-methylpyridine
SMILES CC1=CC(=CN=C1Br)I

Specifications

Melting Point 46°C
Color White-Yellow
Quantity 1 g
Synonym 2-bromo-5-iodo-3-picoline, pyridine, 2-bromo-5-iodo-3-methyl, 2-bromo-5-iodo-3-methyl-pyridine, pubchem5968, acmc-1b7x6, pyridine,2-bromo-5-iodo-3-methyl, 2-bromo-5-iodo-3-picoline; 2-bromo-3-methyl-5-iodopyridine
Formula Weight 297.92
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-iodo-3-methylpyridine