missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-hydroxypyridine 98.0+%, TCI America™

Catalog No. B36575G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36575G 5 g
B36571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36575G Supplier TCI America Supplier No. B36575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 55717-45-8 | C5H6BNO | 106.92 g/mol

Chemical Identifiers

CAS 55717-45-8
Molecular Formula C5H6BNO
Molecular Weight (g/mol) 106.92
MDL Number MFCD04114184
InChI Key ADHMWMAEKRZWPB-UHFFFAOYSA-N
Synonym 2-bromo-5-hydroxypyridine, 6-bromo-3-pyridinol, 6-bromo-3-hydroxypyridine, 3-pyridinol, 6-bromo, 5-hydroxy-2-bromopyridine, pubchem5967, zlchem 1240, 2-bromopyridine-5-ol, acmc-209lor, 3-hydroxy-6-bromopyridine
PubChem CID 2762865
IUPAC Name 6-boranylpyridin-3-ol
SMILES BC1=CC=C(O)C=N1

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 5 g
Formula Weight 174.00
Percent Purity ≥98.0% (GC,T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-hydroxypyridine