missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-hydroxypyridine, 95%

Catalog No. AAH2722806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2722806 5 g
AAH2722803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2722806 Supplier Thermo Scientific Chemicals Supplier No. H2722806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 55717-45-8 | C5H6BNO | 106.92 g/mol

It is used as an active pharmaceutical intermediate and in the coupling reactions with halo aromatic amines and alcohols for a practical synthetic route to 2-substituted amino phenyl and hydroxyphenyl pyridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate and in the coupling reactions with halo aromatic amines and alcohols for a practical synthetic route to 2-substituted amino phenyl and hydroxyphenyl pyridines.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 55717-45-8
Molecular Formula C5H6BNO
Molecular Weight (g/mol) 106.92
MDL Number MFCD04114184
InChI Key ADHMWMAEKRZWPB-UHFFFAOYSA-N
Synonym 2-bromo-5-hydroxypyridine, 6-bromo-3-pyridinol, 6-bromo-3-hydroxypyridine, 3-pyridinol, 6-bromo, 5-hydroxy-2-bromopyridine, pubchem5967, zlchem 1240, 2-bromopyridine-5-ol, acmc-209lor, 3-hydroxy-6-bromopyridine
PubChem CID 2762865
IUPAC Name 6-bromopyridin-3-ol
SMILES BC1=CC=C(O)C=N1

Specifications

Melting Point 133°C to 136°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 174
Percent Purity 95%
Chemical Name or Material 2-Bromo-5-hydroxypyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only