missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-hydroxypyrazine 95.0+%, TCI America™
$256.68 - $256.68
Chemical Identifiers
| CAS | 374063-92-0 |
|---|---|
| Molecular Formula | C4H3BrN2O |
| Molecular Weight (g/mol) | 174.99 |
| MDL Number | MFCD06245330 |
| InChI Key | ITTXBHQAWOFJAI-UHFFFAOYSA-N |
| Synonym | 5-bromopyrazin-2-ol, 2-bromo-5-hydroxypyrazine, 5-bromo-2-pyrazinol, 5-bromo-pyrazin-2-ol, 2 1h-pyrazinone, 5-bromo, 5-bromo-2-hydroxypyrazine, 2-bromo-5-hydroxy-pyrazine, pubchem7870, acmc-20a0of, 2-hydroxy-5-bromopyrazine |
| PubChem CID | 2771618 |
| IUPAC Name | 5-bromo-1,2-dihydropyrazin-2-one |
| SMILES | BrC1=CNC(=O)C=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B3881200MG
|
TCI America
B3881200MG |
200 mg |
Each for $256.68
|
|
|||||
Chemical Identifiers
| 374063-92-0 | |
| 174.99 | |
| ITTXBHQAWOFJAI-UHFFFAOYSA-N | |
| 2771618 | |
| BrC1=CNC(=O)C=N1 |
| C4H3BrN2O | |
| MFCD06245330 | |
| 5-bromopyrazin-2-ol, 2-bromo-5-hydroxypyrazine, 5-bromo-2-pyrazinol, 5-bromo-pyrazin-2-ol, 2 1h-pyrazinone, 5-bromo, 5-bromo-2-hydroxypyrazine, 2-bromo-5-hydroxy-pyrazine, pubchem7870, acmc-20a0of, 2-hydroxy-5-bromopyrazine | |
| 5-bromo-1,2-dihydropyrazin-2-one |
Specifications
| 374063-92-0 | |
| C4H3BrN2O | |
| 200 mg | |
| ITTXBHQAWOFJAI-UHFFFAOYSA-N | |
| 5-bromo-1,2-dihydropyrazin-2-one | |
| 2771618 | |
| ≥95.0% (GC) | |
| 2-Bromo-5-hydroxypyrazine |
| 115°C | |
| MFCD06245330 | |
| 5-bromopyrazin-2-ol, 2-bromo-5-hydroxypyrazine, 5-bromo-2-pyrazinol, 5-bromo-pyrazin-2-ol, 2 1h-pyrazinone, 5-bromo, 5-bromo-2-hydroxypyrazine, 2-bromo-5-hydroxy-pyrazine, pubchem7870, acmc-20a0of, 2-hydroxy-5-bromopyrazine | |
| BrC1=CNC(=O)C=N1 | |
| 174.99 | |
| 174.99 | |
| Crystalline Powder |
Safety and Handling
TSCA : No