Learn More
2-Bromo-5-hydroxypyrazine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 435890010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Bromo-5-hydroxypyrazine | |
| 96.0 | |
| 115°C to 119°C | |
| Authentic | |
| Glass bottle | |
| MFCD06245330 | |
| 5-bromopyrazin-2-ol, 2-bromo-5-hydroxypyrazine, 5-bromo-2-pyrazinol, 5-bromo-pyrazin-2-ol, 2 1h-pyrazinone, 5-bromo, 5-bromo-2-hydroxypyrazine, 2-bromo-5-hydroxy-pyrazine, pubchem7870, acmc-20a0of, 2-hydroxy-5-bromopyrazine | |
| BrC1=CNC(=O)C=N1 | |
| 174.99 | |
| 174.98 | |
| Crystalline Powder |
| 374063-92-0 | |
| 100.0 | |
| White to Brown | |
| 96% min. (HPLC) | |
| C4H3BrN2O | |
| 1 g | |
| ITTXBHQAWOFJAI-UHFFFAOYSA-N | |
| 5-bromo-1,2-dihydropyrazin-2-one | |
| 2771618 | |
| 97% |
Chemical Identifiers
| 374063-92-0 | |
| 174.99 | |
| ITTXBHQAWOFJAI-UHFFFAOYSA-N | |
| 2771618 |
| C4H3BrN2O | |
| MFCD06245330 | |
| 5-bromopyrazin-2-ol, 2-bromo-5-hydroxypyrazine, 5-bromo-2-pyrazinol, 5-bromo-pyrazin-2-ol, 2 1h-pyrazinone, 5-bromo, 5-bromo-2-hydroxypyrazine, 2-bromo-5-hydroxy-pyrazine, pubchem7870, acmc-20a0of, 2-hydroxy-5-bromopyrazine | |
| BrC1=CNC(=O)C=N1 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap
GHS Signal Word: Warning
RUO – Research Use Only