missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-hydroxybenzaldehyde 97.0+%, TCI America™

Catalog No. p-7123292
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39971G 1 g
B39975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39971G Supplier TCI America Supplier No. B39971G
Only null left

Chemical Identifiers

CAS 2973-80-0
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.02
MDL Number MFCD03033065
InChI Key SCRQAWQJSSKCFN-UHFFFAOYSA-N
Synonym 4-bromo-3-formylphenol, 2-bromo-5-hydroxy-benzaldehyde, 2-bromo-5-hydroxy benzaldehyde, benzaldehyde, 2-bromo-5-hydroxy, 2-bromo-5-hydroxybenzadehyde, benzaldehyde,2-bromo-5-hydroxy, pubchem14430, acmc-209haw, 6-brom-3-oxy-benzaldehyd
PubChem CID 387179
IUPAC Name 2-bromo-5-hydroxybenzaldehyde
SMILES OC1=CC(C=O)=C(Br)C=C1

Specifications

Melting Point 132°C
Quantity 1 g
Formula Weight 201.02
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-hydroxybenzaldehyde