missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-hydroxybenzaldehyde, 95%

Catalog No. p-7056125
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6488406 5 g
AAH6488414 25 g
AAH6488422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6488406 Supplier Thermo Scientific Chemicals Supplier No. H6488406
Only null left

CAS: 2973-80-0 | C7H5BrO2 | 201.02 g/mol

2-Bromo-5-hydroxybenzaldehyde is a useful reactant used in the synthesis of PDE4 inhibitors, BCL-XL and cancel cell growth inhibitors. Also an reactant used in the preparation of many pharmaceutical agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-5-hydroxybenzaldehyde is a useful reactant used in the synthesis of PDE4 inhibitors, BCL-XL and cancel cell growth inhibitors. Also an reactant used in the preparation of many pharmaceutical agents.

Solubility
Soluble in chloroform, dichloromethane and ethyl acetate.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 2973-80-0
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.02
MDL Number MFCD03033065
InChI Key SCRQAWQJSSKCFN-UHFFFAOYSA-N
PubChem CID 387179
IUPAC Name 2-bromo-5-hydroxybenzaldehyde
SMILES OC1=CC(C=O)=C(Br)C=C1

Specifications

Melting Point 130°C to 135°C
Quantity 5 g
UN Number UN3077
Beilstein 2079921
Synonym 4-bromo-3-formylphenol, 2-bromo-5-hydroxy-benzaldehyde, 2-bromo-5-hydroxy benzaldehyde, benzaldehyde, 2-bromo-5-hydroxy, 2-bromo-5-hydroxybenzadehyde, benzaldehyde,2-bromo-5-hydroxy, pubchem14430, acmc-209haw, 6-brom-3-oxy-benzaldehyd
Solubility Information Soluble in chloroform,dichloromethane and ethyl acetate.
Formula Weight 201.02
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Bromo-5-hydroxybenzaldehyde
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only