missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluoropyridine 98.0+%, TCI America™

$67.11 - $328.08

Chemical Identifiers

CAS 41404-58-4
Molecular Formula C5H3BrFN
Molecular Weight (g/mol) 175.988
InChI Key UODINHBLNPPDPD-UHFFFAOYSA-N
Synonym 2-bromo-5-fluoro-pyridine, pyridine, 2-bromo-5-fluoro, pubchem1099, pubchem14324, 2-bromo5-fluoropyridine, acmc-1assy, 2-bromo-5-fluoropyridin, 2-bromo-5-fluorpyridine, 2-bromo-5-fluropyridine, 5-fluoro-2-bromopyridine
PubChem CID 2783171
IUPAC Name 2-bromo-5-fluoropyridine
SMILES C1=CC(=NC=C1F)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B27995G
View Documents
TCI America
B27995G
5 g
Each for $67.11
Only null left
 
B279925G
View Documents
TCI America
B279925G
25 g
Each for $328.08
Only null left
 
Description

Description

Specifications

Chemical Identifiers

41404-58-4
175.988
2-bromo-5-fluoro-pyridine, pyridine, 2-bromo-5-fluoro, pubchem1099, pubchem14324, 2-bromo5-fluoropyridine, acmc-1assy, 2-bromo-5-fluoropyridin, 2-bromo-5-fluorpyridine, 2-bromo-5-fluropyridine, 5-fluoro-2-bromopyridine
2-bromo-5-fluoropyridine
C5H3BrFN
UODINHBLNPPDPD-UHFFFAOYSA-N
2783171
C1=CC(=NC=C1F)Br

Specifications

41404-58-4
83°C
5 g
UODINHBLNPPDPD-UHFFFAOYSA-N
2-bromo-5-fluoropyridine
2783171
≥98.0% (GC)
2-Bromo-5-fluoropyridine
White-Yellow
C5H3BrFN
2-bromo-5-fluoro-pyridine, pyridine, 2-bromo-5-fluoro, pubchem1099, pubchem14324, 2-bromo5-fluoropyridine, acmc-1assy, 2-bromo-5-fluoropyridin, 2-bromo-5-fluorpyridine, 2-bromo-5-fluropyridine, 5-fluoro-2-bromopyridine
C1=CC(=NC=C1F)Br
175.988
175.99
Crystals
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'