missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-chlorotoluene 98.0+%, TCI America™
Supplier: TCI America B1427250G
Specifications
| 2-Bromo-5-chlorotoluene | |
| Yellow | |
| 250 g | |
| C7H6BrCl | |
| 2-bromo-5-chlorotoluene, benzene, 1-bromo-4-chloro-2-methyl, toluene, 2-bromo-5-chloro, 6-bromo-3-chlorotoluene, 1-bromo-4-chloro-2-methyl-benzene, 5-chloro-2-bromotoluene, pubchem3600, acmc-209ctq, 2-bromo-5-chloro-toluene, ksc493i7b | |
| CC1=C(C=CC(=C1)Cl)Br | |
| 205.479 | |
| 205.48 |
| 14495-51-3 | |
| 100°C | |
| Clear Liquid at 20°C | |
| MFCD00000602 | |
| RTIPTGMVQIIMKL-UHFFFAOYSA-N | |
| 1-bromo-4-chloro-2-methylbenzene | |
| 84476 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 14495-51-3 | |
| 205.479 | |
| RTIPTGMVQIIMKL-UHFFFAOYSA-N | |
| 84476 | |
| CC1=C(C=CC(=C1)Cl)Br |
| C7H6BrCl | |
| MFCD00000602 | |
| 2-bromo-5-chlorotoluene, benzene, 1-bromo-4-chloro-2-methyl, toluene, 2-bromo-5-chloro, 6-bromo-3-chlorotoluene, 1-bromo-4-chloro-2-methyl-benzene, 5-chloro-2-bromotoluene, pubchem3600, acmc-209ctq, 2-bromo-5-chloro-toluene, ksc493i7b | |
| 1-bromo-4-chloro-2-methylbenzene |
Safety and Handling
TSCA : No