missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-chlorotoluene 98.0+%, TCI America™

Catalog No. B142725G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B142725G 25 g
B1427250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B142725G Supplier TCI America Supplier No. B142725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14495-51-3
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.479
MDL Number MFCD00000602
InChI Key RTIPTGMVQIIMKL-UHFFFAOYSA-N
Synonym 2-bromo-5-chlorotoluene, benzene, 1-bromo-4-chloro-2-methyl, toluene, 2-bromo-5-chloro, 6-bromo-3-chlorotoluene, 1-bromo-4-chloro-2-methyl-benzene, 5-chloro-2-bromotoluene, pubchem3600, acmc-209ctq, 2-bromo-5-chloro-toluene, ksc493i7b
PubChem CID 84476
IUPAC Name 1-bromo-4-chloro-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Cl)Br

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
Boiling Point 100°C
Formula Weight 205.48
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Bromo-5-chlorotoluene