missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-chloropyridine 98.0+%, TCI America™

Catalog No. p-7119751
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27631G 1 g
B27635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27631G Supplier TCI America Supplier No. B27631G
Only null left

Chemical Identifiers

CAS 40473-01-6
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
InChI Key BZUUVQCSPHPUQA-UHFFFAOYSA-N
Synonym 5-chloro-2-bromopyridine, 2-bromo-5-chloro-pyridine, 2-brom-5-chlorpyridin, pyridine, 2-bromo-5-chloro, pubchem1169, acmc-209jeb, 2-bromo-5-chloro pyridine, ksc238i0d, tpc-py102, 2-bromanyl-5-chloranyl-pyridine
PubChem CID 817098
IUPAC Name 2-bromo-5-chloropyridine
SMILES C1=CC(=NC=C1Cl)Br

Specifications

Melting Point 67°C
Boiling Point 128°C
Quantity 1 g
Formula Weight 192.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-chloropyridine