missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-chloropyridine, 98%

Catalog No. AAH6436722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6436722 100 g
AAH6436714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6436722 Supplier Thermo Scientific Chemicals Supplier No. H6436722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 40473-01-6 | C5H3BrClN | 192.44 g/mol

It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is also used in the synthesis of novel halopyridinylboronic acids and esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is also used in the synthesis of novel halopyridinylboronic acids and esters.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 40473-01-6
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
MDL Number MFCD00234006
InChI Key BZUUVQCSPHPUQA-UHFFFAOYSA-N
Synonym 5-chloro-2-bromopyridine, 2-bromo-5-chloro-pyridine, 2-brom-5-chlorpyridin, pyridine, 2-bromo-5-chloro, pubchem1169, acmc-209jeb, 2-bromo-5-chloro pyridine, ksc238i0d, tpc-py102, 2-bromanyl-5-chloranyl-pyridine
PubChem CID 817098
IUPAC Name 2-bromo-5-chloropyridine
SMILES C1=CC(=NC=C1Cl)Br

Specifications

Melting Point 65°C to 69°C (literature)
Boiling Point 128°C (16 mmHg)
Quantity 100 g
Beilstein 109098
Solubility Information Slightly soluble in water.
Formula Weight 192.44
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-5-chloropyridine
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only