missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-chloro-3-nitropyridine 98.0+%, TCI America™
$119.36 - $346.52
Chemical Identifiers
| CAS | 75806-86-9 |
|---|---|
| Molecular Formula | C5H2BrClN2O2 |
| Molecular Weight (g/mol) | 237.437 |
| MDL Number | MFCD07368982 |
| InChI Key | VFMIDVIAQMFGGP-UHFFFAOYSA-N |
| Synonym | 2-bromo-3-nitro-5-chloropyridine, pyridine, 2-bromo-5-chloro-3-nitro, 2-bromo-5-chloro-3-nitro-pyridine, pubchem5951, acmc-1bggp, ksc495s5l, 2-bromo-5-chloro-3-nitro pyridine, 3-nitro-2-bromo-5-chloropyridine, 5-chloro-2-bromo-3-nitropyridine |
| PubChem CID | 280681 |
| IUPAC Name | 2-bromo-5-chloro-3-nitropyridine |
| SMILES | C1=C(C(=NC=C1Cl)Br)[N+](=O)[O-] |
Chemical Identifiers
| 75806-86-9 | |
| 237.437 | |
| VFMIDVIAQMFGGP-UHFFFAOYSA-N | |
| 280681 | |
| C1=C(C(=NC=C1Cl)Br)[N+](=O)[O-] |
| C5H2BrClN2O2 | |
| MFCD07368982 | |
| 2-bromo-3-nitro-5-chloropyridine, pyridine, 2-bromo-5-chloro-3-nitro, 2-bromo-5-chloro-3-nitro-pyridine, pubchem5951, acmc-1bggp, ksc495s5l, 2-bromo-5-chloro-3-nitro pyridine, 3-nitro-2-bromo-5-chloropyridine, 5-chloro-2-bromo-3-nitropyridine | |
| 2-bromo-5-chloro-3-nitropyridine |
Specifications
| 75806-86-9 | |
| Yellow | |
| MFCD07368982 | |
| 2-bromo-3-nitro-5-chloropyridine, pyridine, 2-bromo-5-chloro-3-nitro, 2-bromo-5-chloro-3-nitro-pyridine, pubchem5951, acmc-1bggp, ksc495s5l, 2-bromo-5-chloro-3-nitro pyridine, 3-nitro-2-bromo-5-chloropyridine, 5-chloro-2-bromo-3-nitropyridine | |
| C1=C(C(=NC=C1Cl)Br)[N+](=O)[O-] | |
| 237.437 | |
| 237.44 | |
| Crystalline Powder |
| 77°C | |
| C5H2BrClN2O2 | |
| 5 g | |
| VFMIDVIAQMFGGP-UHFFFAOYSA-N | |
| 2-bromo-5-chloro-3-nitropyridine | |
| 280681 | |
| ≥98.0% (GC) | |
| 2-Bromo-5-chloro-3-nitropyridine |
Safety and Handling
TSCA : No