missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-chloro-3-nitropyridine 98.0+%, TCI America™

Catalog No. B43105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43105G 5 g
B431025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43105G Supplier TCI America Supplier No. B43105G

Chemical Identifiers

CAS 75806-86-9
Molecular Formula C5H2BrClN2O2
Molecular Weight (g/mol) 237.437
MDL Number MFCD07368982
InChI Key VFMIDVIAQMFGGP-UHFFFAOYSA-N
Synonym 2-bromo-3-nitro-5-chloropyridine, pyridine, 2-bromo-5-chloro-3-nitro, 2-bromo-5-chloro-3-nitro-pyridine, pubchem5951, acmc-1bggp, ksc495s5l, 2-bromo-5-chloro-3-nitro pyridine, 3-nitro-2-bromo-5-chloropyridine, 5-chloro-2-bromo-3-nitropyridine
PubChem CID 280681
IUPAC Name 2-bromo-5-chloro-3-nitropyridine
SMILES C1=C(C(=NC=C1Cl)Br)[N+](=O)[O-]

Specifications

Melting Point 77°C
Color Yellow
Quantity 5 g
Formula Weight 237.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-chloro-3-nitropyridine