missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-phenylacetophenone 98.0+%, TCI America™

Catalog No. B06335G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B06335G 5 g
B063325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B06335G Supplier TCI America Supplier No. B06335G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 135-73-9
Molecular Formula C14H11BrO
Molecular Weight (g/mol) 275.145
MDL Number MFCD00000202
InChI Key KGHGZRVXCKCJGX-UHFFFAOYSA-N
Synonym 2-bromo-4'-phenylacetophenone, 4-phenylphenacyl bromide, p-bromoacetylbiphenyl, p-phenylphenacyl bromide, bromomethyl p-biphenylyl ketone, ethanone, 1-1,1'-biphenyl-4-yl-2-bromo, 2-bromo-1-4-phenylphenyl ethan-1-one, acetophenone, 2-bromo-4'-phenyl, alpha-bromo-p-phenylacetophenone, omega-bromo-4-phenylacetophenone
PubChem CID 67282
IUPAC Name 2-bromo-1-(4-phenylphenyl)ethanone
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)CBr

Specifications

Melting Point 125°C
Color Yellow
Quantity 5 g
UN Number 3261
Formula Weight 275.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4′-phenylacetophenone