Learn More
2-Bromo-4'-phenylacetophenone, 97%
CAS: 135-73-9 | C14H11BrO | 275.145 g/mol
Supplier: Thermo Scientific Chemicals B2429814
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Bromo-4'-phenylacetophenone | |
| 123°C to 127°C | |
| MFCD00000202 | |
| UN3261 | |
| Light sensitive | |
| KGHGZRVXCKCJGX-UHFFFAOYSA-N | |
| 2-bromo-1-(4-phenylphenyl)ethanone | |
| 67282 | |
| 97% |
| 135-73-9 | |
| C14H11BrO | |
| 25 g | |
| 513838 | |
| 2-bromo-4'-phenylacetophenone, 4-phenylphenacyl bromide, p-bromoacetylbiphenyl, p-phenylphenacyl bromide, bromomethyl p-biphenylyl ketone, ethanone, 1-1,1'-biphenyl-4-yl-2-bromo, 2-bromo-1-4-phenylphenyl ethan-1-one, acetophenone, 2-bromo-4'-phenyl, alpha-bromo-p-phenylacetophenone, omega-bromo-4-phenylacetophenone | |
| C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)CBr | |
| 275.145 | |
| 275.15 |
Chemical Identifiers
| 135-73-9 | |
| 275.145 | |
| KGHGZRVXCKCJGX-UHFFFAOYSA-N | |
| 67282 | |
| C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)CBr |
| C14H11BrO | |
| MFCD00000202 | |
| 2-bromo-4'-phenylacetophenone, 4-phenylphenacyl bromide, p-bromoacetylbiphenyl, p-phenylphenacyl bromide, bromomethyl p-biphenylyl ketone, ethanone, 1-1,1'-biphenyl-4-yl-2-bromo, 2-bromo-1-4-phenylphenyl ethan-1-one, acetophenone, 2-bromo-4'-phenyl, alpha-bromo-p-phenylacetophenone, omega-bromo-4-phenylacetophenone | |
| 2-bromo-1-(4-phenylphenyl)ethanone |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335-H500
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 205-217-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only