missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4-nitropyridine 95.0+%, TCI America™
$61.51 - $216.24
Chemical Identifiers
| CAS | 6945-67-1 |
|---|---|
| Molecular Formula | C5H3BrN2O2 |
| Molecular Weight (g/mol) | 203.00 |
| MDL Number | MFCD03701383 |
| InChI Key | AFVITJKRFRRQKT-UHFFFAOYSA-N |
| Synonym | pyridine, 2-bromo-4-nitro, 2-brom-4-nitropyridin, pubchem13197, acmc-1badj, 2-bromo-4-nitro-pyridine, 2-bromanyl-4-nitro-pyridine, ksc494c7j, 2-bromo-4-nitropyridine, abbypharma ap-10-5158 |
| PubChem CID | 243015 |
| IUPAC Name | 2-bromo-4-nitropyridine |
| SMILES | [O-][N+](=O)C1=CC=NC(Br)=C1 |
Chemical Identifiers
| 6945-67-1 | |
| 203.00 | |
| AFVITJKRFRRQKT-UHFFFAOYSA-N | |
| 243015 | |
| [O-][N+](=O)C1=CC=NC(Br)=C1 |
| C5H3BrN2O2 | |
| MFCD03701383 | |
| pyridine, 2-bromo-4-nitro, 2-brom-4-nitropyridin, pubchem13197, acmc-1badj, 2-bromo-4-nitro-pyridine, 2-bromanyl-4-nitro-pyridine, ksc494c7j, 2-bromo-4-nitropyridine, abbypharma ap-10-5158 | |
| 2-bromo-4-nitropyridine |
Specifications
| 6945-67-1 | |
| C5H3BrN2O2 | |
| 1 g | |
| AFVITJKRFRRQKT-UHFFFAOYSA-N | |
| 2-bromo-4-nitropyridine | |
| 243015 | |
| ≥95.0% (GC) | |
| 2-Bromo-4-nitropyridine |
| 62°C | |
| MFCD03701383 | |
| pyridine, 2-bromo-4-nitro, 2-brom-4-nitropyridin, pubchem13197, acmc-1badj, 2-bromo-4-nitro-pyridine, 2-bromanyl-4-nitro-pyridine, ksc494c7j, 2-bromo-4-nitropyridine, abbypharma ap-10-5158 | |
| [O-][N+](=O)C1=CC=NC(Br)=C1 | |
| 203.00 | |
| 203.00 | |
| Crystalline Powder |
Safety and Handling
TSCA : No